Anti-MYOD Mouse Monoclonal Antibody [clone: 474515]

Supplier: R & D Systems
MAB5966
220021-303EA 577.02 USD
220021-303
Anti-MYOD Mouse Monoclonal Antibody [clone: 474515]
Antibodies
Type: Primary
Antigen: MYOD
Clonality: Monoclonal
Clone: 474515
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR