Anti-REG3A Mouse Monoclonal Antibody [clone: 512124]

Supplier: R & D Systems
MAB5965
220021-302EA 594.37 USD
220021-302
Anti-REG3A Mouse Monoclonal Antibody [clone: 512124]
Antibodies
Type: Primary
Antigen: REG3A
Clonality: Monoclonal
Clone: 512124
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR