Anti-IL21R Rat Monoclonal Antibody [clone: 155516]

Supplier: R & D Systems
MAB5961
220021-301EA 561.11 USD
220021-301
Anti-IL21R Rat Monoclonal Antibody [clone: 155516]
Antibodies
Type: Primary
Antigen: IL21R
Clonality: Monoclonal
Clone: 155516
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR