Anti-IL21R Rat Monoclonal Antibody [clone: 155502]

Supplier: R & D Systems
MAB596
220021-300EA 763.57 USD
220021-300
Anti-IL21R Rat Monoclonal Antibody [clone: 155502]
Antibodies
Type: Primary
Antigen: IL21R
Clonality: Monoclonal
Clone: 155502
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR