Anti-RPTOR Mouse Monoclonal Antibody [clone: 514208]

Supplier: R & D Systems
MAB5957
102886-154EA 594.37 USD
102886-154
Anti-RPTOR Mouse Monoclonal Antibody [clone: 514208]
Antibodies
Type: Primary
Antigen: RPTOR
Clonality: Monoclonal
Clone: 514208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR