Anti-LRP4 Mouse Monoclonal Antibody [clone: 741704]

Supplier: R & D Systems
MAB5948
102886-148EA 600.15 USD
102886-148
Anti-LRP4 Mouse Monoclonal Antibody [clone: 741704]
Antibodies
Type: Primary
Antigen: LRP4
Clonality: Monoclonal
Clone: 741704
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR