Anti-FMOD Mouse Monoclonal Antibody [clone: 549302]

Supplier: R & D Systems
MAB5945
220021-297EA 594.37 USD
220021-297
Anti-FMOD Mouse Monoclonal Antibody [clone: 549302]
Antibodies
Type: Primary
Antigen Symbol: FMOD
Antigen Name: fibromodulin
Antigen Synonyms: FM, SLRR2E
Clonality: Monoclonal
Clone: 549302
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR