Anti-IL21 Rat Monoclonal Antibody [clone: 149204]

Supplier: R & D Systems
MAB594
220021-296EA 779.48 USD
220021-296
Anti-IL21 Rat Monoclonal Antibody [clone: 149204]
Antibodies
Type: Primary
Antigen: IL21
Clonality: Monoclonal
Clone: 149204
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR