Anti-FOXO1 Mouse Monoclonal Antibody [clone: 597554]

Supplier: R & D Systems
MAB5939
102886-146EA 584.25 USD
102886-146
Anti-FOXO1 Mouse Monoclonal Antibody [clone: 597554]
Antibodies
Type: Primary
Antigen Symbol: FOXO1
Antigen Name: forkhead box O1
Antigen Synonyms: FKH1, FKHR, FOXO1A
Clonality: Monoclonal
Clone: 597554
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR