Anti-FHL1 Mouse Monoclonal Antibody [clone: 767418]

Supplier: R & D Systems
MAB5938
102885-976EA 594.37 USD
102885-976
Anti-FHL1 Mouse Monoclonal Antibody [clone: 767418]
Antibodies
Type: Primary
Antigen Symbol: FHL1
Antigen Name: four and a half LIM domains 1
Antigen Synonyms: FHL-1, FHL1A, FHL1B, FLH1A, KYOT, SLIM, SLIM-1, SLIM1, SLIMMER, XMPMA
Clonality: Monoclonal
Clone: 767418
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR