Anti-ORM1 Rat Monoclonal Antibody [clone: 628110]

Supplier: R & D Systems
MAB5934
220021-295EA 594.37 USD
220021-295
Anti-ORM1 Rat Monoclonal Antibody [clone: 628110]
Antibodies
Type: Primary
Antigen: ORM1
Clonality: Monoclonal
Clone: 628110
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR