Anti-TNFRSF17 Rat Monoclonal Antibody [clone: 161608]

Supplier: R & D Systems
MAB5931
220021-294EA 763.57 USD
220021-294
Anti-TNFRSF17 Rat Monoclonal Antibody [clone: 161608]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: TNFRSF17
Clonality: Monoclonal
Clone: 161608
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR