Anti-TNFRSF17 Rat Monoclonal Antibody [clone: 161616]

Supplier: R & D Systems
MAB593
220021-293EA 763.57 USD
220021-293
Anti-TNFRSF17 Rat Monoclonal Antibody [clone: 161616]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: TNFRSF17
Clonality: Monoclonal
Clone: 161616
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR