Anti-PPP2R4 Mouse Monoclonal Antibody [clone: 529245]

Supplier: R & D Systems
MAB5929
220021-292EA 577.02 USD
220021-292
Anti-PPP2R4 Mouse Monoclonal Antibody [clone: 529245]
Antibodies
Type: Primary
Antigen: PPP2R4
Clonality: Monoclonal
Clone: 529245
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR