Anti-FASN Mouse Monoclonal Antibody [clone: 647114]

Supplier: R & D Systems
MAB59271
102885-972EA 577.02 USD
102885-972
Anti-FASN Mouse Monoclonal Antibody [clone: 647114]
Antibodies
Type: Primary
Antigen Symbol: FASN
Antigen Name: fatty acid synthase
Antigen Synonyms: FAS, OA-519, SDR27X1
Clonality: Monoclonal
Clone: 647114
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR