Anti-FASN Mouse Monoclonal Antibody [clone: 647113]

Supplier: R & D Systems
MAB5927
102885-970EA 594.37 USD
102885-970
Anti-FASN Mouse Monoclonal Antibody [clone: 647113]
Antibodies
Type: Primary
Antigen Symbol: FASN
Antigen Name: fatty acid synthase
Antigen Synonyms: A630082H08Rik, FAS
Clonality: Monoclonal
Clone: 647113
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR