Anti-ST6GAL1 Mouse Monoclonal Antibody [clone: 396012]

Supplier: R & D Systems
MAB5924
102885-968EA 600.15 USD
102885-968
Anti-ST6GAL1 Mouse Monoclonal Antibody [clone: 396012]
Antibodies
Type: Primary
Antigen: ST6GAL1
Clonality: Monoclonal
Clone: 396012
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR