Anti-SLC3A2 Mouse Monoclonal Antibody [clone: 590559]

Supplier: R & D Systems
MAB5920
102885-966EA 600.15 USD
102885-966
Anti-SLC3A2 Mouse Monoclonal Antibody [clone: 590559]
Antibodies
Type: Primary
Antigen: SLC3A2
Clonality: Monoclonal
Clone: 590559
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR