Anti-BMP15 Rat Monoclonal Antibody [clone: 582703]

Supplier: R & D Systems
MAB5917
220021-291EA 561.11 USD
220021-291
Anti-BMP15 Rat Monoclonal Antibody [clone: 582703]
Antibodies
Type: Primary
Antigen: BMP15
Clonality: Monoclonal
Clone: 582703
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR