Anti-ADGRL3 Mouse Monoclonal Antibody [clone: 81518]

Supplier: R & D Systems
MAB5916
102885-964EA 577.02 USD
102885-964
Anti-ADGRL3 Mouse Monoclonal Antibody [clone: 81518]
Antibodies
Simple Western

Type: Primary
Antigen: ADGRL3
Clonality: Monoclonal
Clone: 81518
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR