Anti-DMBT1 Rat Monoclonal Antibody [clone: 548012]

Supplier: R & D Systems
MAB59151
102885-962EA 577.02 USD
102885-962
Anti-DMBT1 Rat Monoclonal Antibody [clone: 548012]
Antibodies
Type: Primary
Antigen: DMBT1
Clonality: Monoclonal
Clone: 548012
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR