Anti-PRKACA Mouse Monoclonal Antibody [clone: 515741]

Supplier: R & D Systems
MAB5908
220021-288EA 577.02 USD
220021-288
Anti-PRKACA Mouse Monoclonal Antibody [clone: 515741]
Antibodies
Type: Primary
Antigen: PRKACA
Clonality: Monoclonal
Clone: 515741
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR