Anti-ALPP Mouse Monoclonal Antibody [clone: 696128]

Supplier: R & D Systems
MAB59051
102885-960EA 600.15 USD
102885-960
Anti-ALPP Mouse Monoclonal Antibody [clone: 696128]
Antibodies
Simple Western

Type: Primary
Antigen: ALPP
Clonality: Monoclonal
Clone: 696128
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR