Anti-ALPP Mouse Monoclonal Antibody [clone: 69618]

Supplier: R & D Systems
MAB5905
102885-958EA 594.37 USD
102885-958
Anti-ALPP Mouse Monoclonal Antibody [clone: 69618]
Antibodies
Type: Primary
Antigen: ALPP
Clonality: Monoclonal
Clone: 69618
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR