Anti-STRA6 Mouse Monoclonal Antibody [clone: 496613]

Supplier: R & D Systems
MAB5904
220021-287EA 594.37 USD
220021-287
Anti-STRA6 Mouse Monoclonal Antibody [clone: 496613]
Antibodies
Type: Primary
Antigen: STRA6
Clonality: Monoclonal
Clone: 496613
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR