Anti-CCR6 Rat Monoclonal Antibody [clone: 140706]

Supplier: R & D Systems
MAB590-100
102885-956EA 633.42 USD
102885-956
Anti-CCR6 Rat Monoclonal Antibody [clone: 140706]
Antibodies
Type: Primary
Antigen: CCR6
Clonality: Monoclonal
Clone: 140706
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR