Anti-PCDH1 Mouse Monoclonal Antibody [clone: 648127]

Supplier: R & D Systems
MAB5899
102885-954EA 561.11 USD
102885-954
Anti-PCDH1 Mouse Monoclonal Antibody [clone: 648127]
Antibodies
Type: Primary
Antigen: PCDH1
Clonality: Monoclonal
Clone: 648127
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR