Anti-PGR Mouse Monoclonal Antibody [clone: 569012]

Supplier: R & D Systems
MAB5886
220021-283EA 594.37 USD
220021-283
Anti-PGR Mouse Monoclonal Antibody [clone: 569012]
Antibodies
Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 569012
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR