Anti-DCC Mouse Monoclonal Antibody [clone: 531505]

Supplier: R & D Systems
MAB5884
220021-281EA 594.37 USD
220021-281
Anti-DCC Mouse Monoclonal Antibody [clone: 531505]
Antibodies
Type: Primary
Antigen: DCC
Clonality: Monoclonal
Clone: 531505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR