Anti-AR Mouse Monoclonal Antibody [clone: 636005]

Supplier: R & D Systems
MAB58761
102885-952EA 577.02 USD
102885-952
Anti-AR Mouse Monoclonal Antibody [clone: 636005]
Antibodies
Type: Primary
Antigen: AR
Clonality: Monoclonal
Clone: 636005
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR