Anti-AR Mouse Monoclonal Antibody [clone: 523339]

Supplier: R & D Systems
MAB5876
220021-280EA 594.37 USD
220021-280
Anti-AR Mouse Monoclonal Antibody [clone: 523339]
Antibodies
Type: Primary
Antigen: AR
Clonality: Monoclonal
Clone: 523339
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR