Anti-SPI1 Mouse Monoclonal Antibody [clone: 732322]

Supplier: R & D Systems
MAB5870
102885-950EA 594.37 USD
102885-950
Anti-SPI1 Mouse Monoclonal Antibody [clone: 732322]
Antibodies
Type: Primary
Antigen: SPI1
Clonality: Monoclonal
Clone: 732322
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR