Anti-TGFBR1 Rat Monoclonal Antibody [clone: 141229]

Supplier: R & D Systems
MAB587
220021-277EA 772.25 USD
220021-277
Anti-TGFBR1 Rat Monoclonal Antibody [clone: 141229]
Antibodies
Type: Primary
Antigen: TGFBR1
Clonality: Monoclonal
Clone: 141229
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR