Anti-ALDH1A1 Mouse Monoclonal Antibody [clone: 7341]

Supplier: R & D Systems
MAB5869
102885-948EA 633.42 USD
102885-948
Anti-ALDH1A1 Mouse Monoclonal Antibody [clone: 7341]
Antibodies
Simple Western

Type: Primary
Antigen: ALDH1A1
Clonality: Monoclonal
Clone: 7341
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR