Anti-ARG1 Mouse Monoclonal Antibody [clone: 658922]

Supplier: R & D Systems
MAB58681
102885-946EA 633.42 USD
102885-946
Anti-ARG1 Mouse Monoclonal Antibody [clone: 658922]
Antibodies
Type: Primary
Antigen: ARG1
Clonality: Monoclonal
Clone: 658922
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR