Anti-ARG1 Mouse Monoclonal Antibody [clone: 658934]

Supplier: R & D Systems
MAB5868
102886-144EA 633.42 USD
102886-144
Anti-ARG1 Mouse Monoclonal Antibody [clone: 658934]
Antibodies
Simple Western

Type: Primary
Antigen: ARG1
Clonality: Monoclonal
Clone: 658934
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR