Anti-PMP2 Mouse Monoclonal Antibody [clone: 631306]

Supplier: R & D Systems
MAB5866
102886-142EA 561.11 USD
102886-142
Anti-PMP2 Mouse Monoclonal Antibody [clone: 631306]
Antibodies
Type: Primary
Antigen: PMP2
Clonality: Monoclonal
Clone: 631306
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR