Anti-NMNAT1 Mouse Monoclonal Antibody [clone: 675112]

Supplier: R & D Systems
MAB5865
102886-140EA 577.02 USD
102886-140
Anti-NMNAT1 Mouse Monoclonal Antibody [clone: 675112]
Antibodies
Type: Primary
Antigen: NMNAT1
Clonality: Monoclonal
Clone: 675112
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR