Anti-TNFSF13 Mouse Monoclonal Antibody [clone: 670820]

Supplier: R & D Systems
MAB5860
102886-138EA 772.25 USD
102886-138
Anti-TNFSF13 Mouse Monoclonal Antibody [clone: 670820]
Antibodies
Type: Primary
Antigen: TNFSF13
Clonality: Monoclonal
Clone: 670820
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR