Anti-NKX6-1 Mouse Monoclonal Antibody [clone: 631438]

Supplier: R & D Systems
MAB5857
102886-136EA 616.06 USD
102886-136
Anti-NKX6-1 Mouse Monoclonal Antibody [clone: 631438]
Antibodies
Type: Primary
Antigen Symbol: NKX6-1
Antigen Name: NK6 homeobox 1
Antigen Synonyms: NKX6.1, NKX6A
Clonality: Monoclonal
Clone: 631438
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR