Anti-PLXNA4 Mouse Monoclonal Antibody [clone: 707201]

Supplier: R & D Systems
MAB5856
102885-324EA 594.37 USD
102885-324
Anti-PLXNA4 Mouse Monoclonal Antibody [clone: 707201]
Antibodies
Type: Primary
Antigen: PLXNA4
Clonality: Monoclonal
Clone: 707201
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR