Anti-VAPB Mouse Monoclonal Antibody [clone: 736904]

Supplier: R & D Systems
MAB58551
102885-322EA 577.02 USD
102885-322
Anti-VAPB Mouse Monoclonal Antibody [clone: 736904]
Antibodies
Type: Primary
Antigen: VAPB
Clonality: Monoclonal
Clone: 736904
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR