Anti-VAPB Mouse Monoclonal Antibody [clone: 695791]

Supplier: R & D Systems
MAB5855
102885-320EA 561.11 USD
102885-320
Anti-VAPB Mouse Monoclonal Antibody [clone: 695791]
Antibodies
Type: Primary
Antigen: VAPB
Clonality: Monoclonal
Clone: 695791
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR