Anti-GAL Mouse Monoclonal Antibody [clone: 581403]

Supplier: R & D Systems
MAB5854
102886-134EA 633.42 USD
102886-134
Anti-GAL Mouse Monoclonal Antibody [clone: 581403]
Antibodies
Type: Primary
Antigen Symbol: GAL
Antigen Name: galanin/GMAP prepropeptide
Antigen Synonyms: GAL-GMAP, GALN, GLNN, GMAP
Clonality: Monoclonal
Clone: 581403
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR