Anti-IL17A Mouse Monoclonal Antibody [clone: 665909]

Supplier: R & D Systems
MAB5848
102886-132EA 763.57 USD
102886-132
Anti-IL17A Mouse Monoclonal Antibody [clone: 665909]
Antibodies
Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: 665909
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR