Anti-FGF4 Rat Monoclonal Antibody [clone: 678431]

Supplier: R & D Systems
MAB5846
102886-128EA 566.89 USD
102886-128
Anti-FGF4 Rat Monoclonal Antibody [clone: 678431]
Antibodies
Type: Primary
Antigen Symbol: FGF4
Antigen Name: fibroblast growth factor 4
Antigen Synonyms: Fgf-4, Fgfk, Hst1, Hstf-1, KS3, hst, hst-1, kFGF
Clonality: Monoclonal
Clone: 678431
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR