Anti-IL4 Mouse Monoclonal Antibody [clone: 143514]

Supplier: R & D Systems
MAB584
220021-272EA 763.57 USD
220021-272
Anti-IL4 Mouse Monoclonal Antibody [clone: 143514]
Antibodies
Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: 143514
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cotton Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR