Anti-VRK1 Mouse Monoclonal Antibody [clone: 1F6]

Supplier: R & D Systems
MAB5835
220021-271EA 594.37 USD
220021-271
Anti-VRK1 Mouse Monoclonal Antibody [clone: 1F6]
Antibodies
Type: Primary
Antigen: VRK1
Clonality: Monoclonal
Clone: 1F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR