Anti-NFATC3 Mouse Monoclonal Antibody [clone: 646115]

Supplier: R & D Systems
MAB5834
102886-126EA 561.11 USD
102886-126
Anti-NFATC3 Mouse Monoclonal Antibody [clone: 646115]
Antibodies
Type: Primary
Antigen Symbol: NFATC3
Antigen Name: nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 3
Antigen Synonyms: NFAT4, NFATX
Clonality: Monoclonal
Clone: 646115
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR