Anti-ICAM1 Mouse Monoclonal Antibody [clone: 141011]

Supplier: R & D Systems
MAB583
220021-268EA 763.57 USD
220021-268
Anti-ICAM1 Mouse Monoclonal Antibody [clone: 141011]
Antibodies
Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: 141011
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR